
α,2,6-Trichlorotoluene, Sigma-Aldrich®
Catalog # SIALT56308-25G
Supplier: Merck
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:25 g
- Seq:3308
Specifications
About this item
:
:195.47 g/mol
Boiling Pt:248 °C (1013 hPa)
Melting Pt:49…50 °C
ADR:8,II
:M-Clarity™ quality level = MQ100
EINECS:217-940-4
UN:3261
:GHS05
CAS Number:2014-83-7
Formula:C₇H₅Cl₃
MDL Number:MFCD00000897