Order Entry
Export
ContactUsLinkComponent
α,2,6-Trichlorotoluene
Corrosive
α,2,6-Trichlorotoluene
Catalog # EHERC17787600
Supplier:  EHRENSTORFER
CAS Number:  
Corrosive
α,2,6-Trichlorotoluene
Catalog # EHERC17787600
Supplier:  EHRENSTORFER
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    0,25 g
  • Seq:
    3029

Specifications

About this item

 :
 :195.47 g/mol
Boiling Pt:248 °C (1013 hPa)
Melting Pt:49…50 °C
EINECS:217-940-4
UN:3261
ADR:8,II
 :GHS05
CAS Number:2014-83-7
Formula:C₇H₅Cl₃
MDL Number:MFCD00000897