
α,2,4-Trichlorotoluene, Sigma-Aldrich®
Catalog # SIAL139254-100ML
Supplier: Merck
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:100 mL
- Seq:2440
Specifications
About this item
:
:195.47 g/mol
Boiling Pt:248 °C (1013 hPa)
Melting Pt:–2.7 °C
Density:1.406 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:202-381-0
UN:3265
ADR:8,II
:M-Clarity™ quality level = MQ100
:GHS05
CAS Number:94-99-5
Formula:C₇H₅Cl₃
MDL Number:MFCD00000895