
2,6-Dichlorotoluene, Sigma-Aldrich®
Catalog # SIALD76005-100G
Supplier: Merck
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:100 g
- Seq:3401
Specifications
About this item
:
:161.03 g/mol
Boiling Pt:200 °C (1013 hPa)
Melting Pt:2 °C
Density:1.2686 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:204-269-7
UN:3082
ADR:9,III
:M-Clarity™ quality level = MQ100
:GHS09
CAS Number:118-69-4
Formula:C₇H₆Cl₂
MDL Number:MFCD00000576