Order Entry
Export
Orders LinkContactUsLinkComponent
ω-Bromo-4-fluoroacetophenone ≥97%

ω-Bromo-4-fluoroacetophenone ≥97%

ω-Bromo-4-fluoroacetophenone ≥97%
Certificates
ω-Bromo-4-fluoroacetophenone ≥97%
ω-Bromo-4-fluoroacetophenone ≥97%

3 Options Available Below

Product Details & Documents

Purity: 97% CAS No.: 403-29-2 Molecular Formula: C8H6BrFO Formula Weight: 217.04 Storage Temperature: 2°C to 8°C Flash Point: 110°(230°F) Melting Point: 45-49° Boiling Point: 150°/12mm Beilstein Reference: 637863 MDL No.: MFCD00040830

Synonyms:  Bromomethyl p-fluorophenyl ketone, α-Bromo-4-fluoroacetophenone, 2-Bromo-1-(4-fluorophenyl)ethan-1-one, 2-Bromo-4'-fluoroacetophenone, 4-Fluorophenacyl bromide, 2-Bromo-1-(4-fluorophenyl)ethanone
  • CAS Number:403-29-2
  • Formula:C₈H₆BrFO
  • MDL Number:MFCD00040830
  • Molecular Weight:217.04 g/mol
  • Boiling Pt:150 °C (12 mmHg)
  • Melting Pt:45…49 °C
  • Flash Pt:>110 °C (230 °F)
  • EINECS:206-955-1
Product Family Options

Product Information