Order Entry
Export
Orders LinkContactUsLinkComponent
 

ω-Bromo-4-fluoroacetophenone 98%

Corrosive
ω-Bromo-4-fluoroacetophenone 98%
Certificates
ω-Bromo-4-fluoroacetophenone 98%

2 Options Available Below

Product Details & Documents
Synonyms:  Bromomethyl p-fluorophenyl ketone, α-Bromo-4-fluoroacetophenone, 2-Bromo-1-(4-fluorophenyl)ethan-1-one, 2-Bromo-4'-fluoroacetophenone, 4-Fluorophenacyl bromide, 2-Bromo-1-(4-fluorophenyl)ethanone
  • CAS Number:403-29-2
  • Formula:C₈H₆BrFO
  • MDL Number:MFCD00040830
  • Molecular Weight:217.04 g/mol
  • Boiling Pt:150 °C (12 mmHg)
  • Melting Pt:46…49 °C
  • Flash Pt:>110 °C
  • Storage Temperature:Refrigerator
  • EINECS:206-955-1
  • UN:3261
  • ADR:8,III

 

Specifications for Product Family

Specification Test Results

  • Appearance:
    White to yellow to yellow-green to beige Powder or crystals and/or chunks
  • Infrared spectrum:
    Conforms
  • Melting point:
    45 to 50 °C
  • GC:
    ≥96.0 %
Product Family Options

Product Information

  • Pack type
    Pk
    Availability
    Price
  • Glass bottle for solids
    5 g
    Log in for availability
    Log in for availability
    1 * 5 g
    1 * 5 g - 52,10 €
    52,10 €
    1 * 5 g
       52,10 €

  • Glass bottle
    25 g
    Log in for availability
    Log in for availability
    1 * 25 g
    1 * 25 g - 170,00 €
    170,00 €
    1 * 25 g
       170,00 €