
α-Bromo-2,6-difluorotoluene, Sigma-Aldrich®
Supplier: Merck
Certificates
About this item
CAS Number:85118-00-9
:
Formula:C₇H₅BrF₂
MDL Number:MFCD00000329
Melting Pt:53 °C
Density:1.6 g/cm³ (20 °C)
Storage Temperature:Ambient
ADR:8,II
:M-Clarity™ quality level = MQ200
:207.02 g/mol
EINECS:285-652-6
UN:3261
:GHS05