2,5-Dibromo-α,α,α-trifluorotoluene, Sigma-Aldrich®
Catalog # SIAL540277-5G
Supplier: Merck
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:5 g
- Seq:3173
Specifications
About this item
:
:303.9 g/mol
:M-Clarity™ quality level = MQ100
EINECS:231-620-1
CAS Number:7657-09-2
Formula:C₇H₃Br₂F₃
MDL Number:MFCD00013554