(4-Bromophenyl)acetonitrile ≥97%
Supplier: Thermo Fisher Scientific
Jump to:
About this item
Purity: 97% CAS No.: 16532-79-9 Molecular Formula: C8H6BrN Formula Weight: 196.05 Flash Point: 127°(261°F) Odour: Pungent Boiling Point: 286° Beilstein Reference: 1861069 UN No.: 3449 MDL No.: MFCD00001916
Synonyms: 4-Bromophenylacetonitrile, 4-Bromobenzyl cyanide
- CAS Number:16532-79-9
- Formula:C₈H₆BrN
- MDL Number:MFCD00001916
- Molecular Weight:196.05 g/mol
- Boiling Pt:286 °C
- Melting Pt:45…49 °C
- Density:1.49
- Flash Pt:127 °C (261 °F)
- Storage Temperature:Ambient
- EINECS:240-602-2
- UN:3449
- ADR:6.1,I
2 Options Available Below
Product Details & Documents
Purity: 97% CAS No.: 16532-79-9 Molecular Formula: C8H6BrN Formula Weight: 196.05 Flash Point: 127°(261°F) Odour: Pungent Boiling Point: 286° Beilstein Reference: 1861069 UN No.: 3449 MDL No.: MFCD00001916
Synonyms: 4-Bromophenylacetonitrile, 4-Bromobenzyl cyanide
- CAS Number:16532-79-9
- Formula:C₈H₆BrN
- MDL Number:MFCD00001916
- Molecular Weight:196.05 g/mol
- Boiling Pt:286 °C
- Melting Pt:45…49 °C
- Density:1.49
- Flash Pt:127 °C (261 °F)
- Storage Temperature:Ambient
- EINECS:240-602-2
- UN:3449
- ADR:6.1,I



