(4-Bromophenyl)acetonitrile 99%
Supplier: Thermo Fisher Scientific
About this item
Synonyms: 4-Bromophenylacetonitrile, 4-Bromobenzyl cyanide
- CAS Number:16532-79-9
- Formula:C₈H₆BrN
- MDL Number:MFCD00001916
- Molecular Weight:196.05 g/mol
- Boiling Pt:286 °C (1013 hPa)
- Melting Pt:47…49 °C
- Flash Pt:152 °C
- EINECS:240-602-2
- UN:3439
- ADR:6.1,III
2 Options Available Below
Product Details & Documents
Synonyms: 4-Bromophenylacetonitrile, 4-Bromobenzyl cyanide
- CAS Number:16532-79-9
- Formula:C₈H₆BrN
- MDL Number:MFCD00001916
- Molecular Weight:196.05 g/mol
- Boiling Pt:286 °C (1013 hPa)
- Melting Pt:47…49 °C
- Flash Pt:152 °C
- EINECS:240-602-2
- UN:3439
- ADR:6.1,III
Documents
ACRO15448
SDS
Specifications for Product Family
Specification Test Results
- Appearance:Colorless to light brown Crystalline mass
- Infrared spectrum:Conforms
- Melting point:45 to 49 °C
- GC:≥98.5 %



