ω-Bromo-3,4-dichloroacetophenone
Catalog # MOLE44437650-5G
Supplier: Molekula
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:5 g
- Seq:3173
Specifications
About this item
:
:267.94 g/mol
Storage Temperature:Refrigerator
CAS Number:2632-10-2
Formula:C₈H₅BrCl₂O
MDL Number:MFCD00051581