
α-Bromo-3,4-dichlorotoluene, Sigma-Aldrich®
Catalog # SIAL681997-1G
Supplier: Merck
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:239.93 g/mol
Boiling Pt:84…86 °C (0.3 mmHg)
ADR:8,II
:M-Clarity™ quality level = MQ100
UN:3265
:GHS05
CAS Number:18880-04-1
Formula:C₇H₅BrCl₂
MDL Number:MFCD00041363