
α-Bromo-3,4-dichlorotoluene 97%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:18880-04-1
:
Formula:C₇H₅BrCl₂
MDL Number:MFCD00041363
:239.93 g/mol
Boiling Pt:115…120 °C (1 mmHg)
Density:1.65 g/cm³
Storage Temperature:Ambient
UN:3265
ADR:8,III
:GHS05
Specification Test Results
- Appearance:Clear colorless to brown Liquid
- Infrared spectrum:Conforms
- Refractive index:1.6050 to 1.6080 (20°C, 589 nm)
- GC:≥96.0 %