
α-Bromo-2,3-dichlorotoluene 99%
Catalog # ACRO365580050
Supplier: Thermo Fisher Scientific
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:5 g
- Seq:3173
Specifications
Specification Test Results
- Appearance:White Low melting solid
- Infrared spectrum:Conforms
- Melting point:29 to 32 °C
- GC:≥98.5 %
Specification Test Results
About this item
:
:239.93 g/mol
Boiling Pt:140…144 °C (31 mmHg)
Melting Pt:29…32 °C
UN:3261
ADR:8,III
:GHS05
CAS Number:57915-78-3
Formula:C₇H₅BrCl₂
MDL Number:MFCD03701317