3-Formylphenylboronic acid 97%
Supplier: Thermo Scientific
Certificates
About this item
CAS Number:87199-16-4
:
Formula:C₇H₇BO₃
MDL Number:MFCD00161356
:149.94 g/mol
Melting Pt:100 °C
:GHS07
Storage Temperature:Refrigerator
Specification Test Results
- Appearance:Off-white to beige to light orange Powder or crystals and/or chunks
- Infrared spectrum:Conforms
- HPLC:≥96.0 %