1,4-Divinylbenzene, Sigma-Aldrich®
Catalog # SIAL535583-5ML
Supplier: Merck
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:5 mL
- Seq:2210
Specifications
About this item
:
Boiling Pt:212 °C (1013 hPa)
Melting Pt:30 °C
Density:0.914 g/cm³ (25 °C)
:M-Clarity™ quality level = MQ200
:130.19 g/mol
Storage Temperature:Refrigerator
EINECS:203-266-8
CAS Number:105-06-6
Formula:C₁₀H₁₀
MDL Number:MFCD00008622