Order Entry
Export
Orders LinkContactUsLinkComponent
Resveratrol

Resveratrol

Glass bottle for solids, 1 g
Catalog # MOLEM57735199
Supplier:   Molekula
Resveratrol
Catalog # MOLEM57735199
Supplier:  Molekula
CAS Number:  501-36-0
Resveratrol

Glass bottle for solids, 1 g

CAS Number:  501-36-0
Price
Inquire For Price
Log in for availability
1 * 1 g
Product Details & Documents
Synonyms:  trans-3,4,5-Trihydroxystilbene, (E)-5-(4-Hydroxystyryl)benzene-1,3-diol
  • CAS Number:501-36-0
  • Formula:C₁₄H₁₂O₃
  • MDL Number:MFCD00133799
  • Molecular Weight:228.25 g/mol
  • Merck Index:12,08326
  • Melting Pt:260 °C
  • Density:1.36 g/cm³ (20 °C)
  • Storage Temperature:Freezer
Specifications

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    1 g

Specifications