
1,4-Diphenylbutadiyne ≥99%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:886-66-8
:
Formula:(C₆H₅)CCCC(C₆H₅)
MDL Number:MFCD00004787
:202.26 g/mol
Boiling Pt:338 °C (1013 hPa)
Melting Pt:87 °C
Density:1.163 g/cm³ (20 °C)
EINECS:212-953-1
:GHS07