Benzene, Sigma-Aldrich®
: Merck
:71-43-2
:
:C₆H₆
:MFCD00003009
:78.11 g/mol
:13,01066
:80.1 °C (1013 hPa)
:5.5 °C
:0.878 g/cm³ (20 °C)
:200-753-7
:1114
:3,II
:M-Clarity™ quality level = MQ200
:–11 °C
:01-2119447106-44
:GHS02
:GHS08
:Ambient