
1,3-Benzenedisulphonyl chloride ≥98%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:585-47-7
:
Formula:C₆H₄Cl₂O₄S₂
MDL Number:MFCD00041879
Boiling Pt:145 °C (1 mmHg)
Melting Pt:58…60 °C
Storage Temperature:Ambient
EINECS:209-556-0
UN:3261
ADR:8,II
:275.13 g/mol
:GHS05