Order Entry
Export
Orders LinkContactUsLinkComponent
1,3-Benzenedithiol ≥97%

1,3-Benzenedithiol ≥97%

Corrosive
Irritant
1,3-Benzenedithiol ≥97%
Certificates
1,3-Benzenedithiol ≥97%
1,3-Benzenedithiol ≥97%

2 Options Available Below

Product Details & Documents

Purity: 97% CAS No.: 626-04-0 Molecular Formula: C6H6S2 Formula Weight: 142.24 Storage Temperature: 2°C to 8°C Flash Point: 112°(234°F) Refractive Index: 1.6625 Odour: Stench Boiling Point: 244° Beilstein Reference: 1634412 MDL No.: MFCD00004842

Synonyms:  Benzene-1,3-dithiol
  • CAS Number:626-04-0
  • Formula:C₆H₆S₂
  • MDL Number:MFCD00004842
  • Molecular Weight:142.25 g/mol
  • Boiling Pt:244 °C
  • Melting Pt:27 °C
  • Density:1.24
  • Flash Pt:>112 °C (234 °F)
  • EINECS:210-925-3
  • UN:3335
  • ADR:9,
Product Family Options

Product Information