1,2,4-Trihydroxybenzene, Sigma-Aldrich®
Supplier: Merck
Certificates
About this item
CAS Number:533-73-3
:
Formula:C₆H₆O₃
MDL Number:MFCD00002198
:126.11 g/mol
Merck Index:13,01073
Boiling Pt:362 °C (1013 hPa)
Melting Pt:140 °C
Density:1.45 g/cm³ (20 °C)
:M-Clarity™ quality level = MQ100
EINECS:208-575-1