Pyrocatechol, Sigma-Aldrich®
Supplier: Merck
Jump to:
About this item
Synonyms: o-Dihydroxybenzene, Catechol, 1,2-Dihydroxybenzene
- CAS Number:120-80-9
- Formula:C₆H₆O₂
- MDL Number:MFCD00002188
- Molecular Weight:110.11 g/mol
- Merck Index:13,08089
- Boiling Pt:245 °C (1013 hPa)
- Melting Pt:103…105 °C
- Density:1.0548 g/cm³ (20 °C)
- Flash Pt:127 °C
- Storage Temperature:Ambient
- UN:2811
- ADR:6.1,III
- :M-Clarity™ quality level = MQ100
- EINECS:204-427-5
2 Options Available Below
Product Details & Documents
Synonyms: o-Dihydroxybenzene, Catechol, 1,2-Dihydroxybenzene
- CAS Number:120-80-9
- Formula:C₆H₆O₂
- MDL Number:MFCD00002188
- Molecular Weight:110.11 g/mol
- Merck Index:13,08089
- Boiling Pt:245 °C (1013 hPa)
- Melting Pt:103…105 °C
- Density:1.0548 g/cm³ (20 °C)
- Flash Pt:127 °C
- Storage Temperature:Ambient
- UN:2811
- ADR:6.1,III
- :M-Clarity™ quality level = MQ100
- EINECS:204-427-5



