
3,4-Dimethoxythiophenol, Sigma-Aldrich®
Catalog # SIAL662518-10G
Supplier: Merck
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:10 g
- Seq:3236
Specifications
About this item
:
:170.23 g/mol
Boiling Pt:86…88 °C (0.4 mmHg)
Density:1.19 g/cm³ (20 °C)
ADR:9,
:M-Clarity™ quality level = MQ100
UN:3334
:GHS07
CAS Number:700-96-9
:19689-66-8
Formula:C₈H₁₀O₂S
MDL Number:MFCD00052823