
3,4-Dimethoxyphenol, Sigma-Aldrich®
Catalog # SIAL194468-10G
Supplier: Merck
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:10 g
- Seq:3236
Specifications
About this item
:
:154.17 g/mol
Boiling Pt:120…122 °C (0.2 mmHg)
Melting Pt:83 °C
EINECS:217-995-4
:GHS07
Storage Temperature:Ambient
CAS Number:2033-89-8
Formula:C₈H₁₀O₃
MDL Number:MFCD00008390