1,4-Dimethoxybenzene
Catalog # MOLE12664681-1KG
Supplier: Molekula
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:1 kg
- Seq:4024
Specifications
About this item
:
:138.17 g/mol
Boiling Pt:212.6 °C (1013 hPa)
Melting Pt:54…56 °C
Density:0.874 g/cm³ (24.9 °C)
Storage Temperature:Ambient
EINECS:205-771-9
CAS Number:150-78-7
Formula:C₈H₁₀O₂
MDL Number:MFCD00008401