Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
1,3-Dimethoxy-2-nitrobenzene
1,3-Dimethoxy-2-nitrobenzene
1,3-Dimethoxy-2-nitrobenzene
1,3-Dimethoxy-2-nitrobenzene
Supplier:
Thermo Scientific
Certificates
Expand image
1,3-Dimethoxy-2-nitrobenzene
Supplier:
Thermo Scientific
About this item
CAS Number:
6665-97-0
:
Formula:
C₈H₉NO₄
MDL Number:
MFCD00474587
:
183.16 g/mol