


1,4-Dinitrobenzene, Sigma-Aldrich®
Catalog # SIAL102369-5G
Supplier: Merck
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:5 g
- Seq:3173
Specifications
About this item
:
:168.11 g/mol
Merck Index:13,03301
Boiling Pt:299 °C (1013 hPa)
Melting Pt:172 °C
Density:1.625 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:202-833-7
UN:3443
ADR:6.1,II
:M-Clarity™ quality level = MQ100
:GHS06
:GHS08
:GHS09
CAS Number:100-25-4
Formula:C₆H₄N₂O₄
MDL Number:MFCD00007314