Sulphanilamide ≥99%, AnalaR NORMAPUR® analytical reagent
Catalog # 21159.181
Supplier: VWR Chemicals
CAS Number:
Specifications
- Pack type:Plastic bottle for solids
- Pk:100 g
- Seq:3401
Specifications
Specification Test Results
- Assay (calculated on anhydrous):Min. 99 %
- Insolubility in acid medium:Passes test
- Insolubility in alkaline medium:Passes test
- Melting point:163 - 166 °C
- Heavy metals (as Pb):Max. 20 ppm
- Ignition residue (SO₄):Max. 0.1 %
- Loss on drying (105°C):Max. 0.5 %
- SO₄ (Sulphate):Max. 0.02 %
- SO4 (Sulphate):Max. 0.02 %
Specification Test Results
About this item
:
:172.21 g/mol
Merck Index:13,09010
Boiling Pt:401 °C (1013 hPa)
Melting Pt:163…167 °C
Density:1.46 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:200-563-4
CAS Number:63-74-1
Formula:H₂NC₆H₄SO₂NH₂
MDL Number:MFCD00007939