Sulphanilamide
Catalog # EHERC17000000
Supplier: EHRENSTORFER
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:0,25 g
- Seq:3029
Specifications
About this item
:
:172.21 g/mol
Merck Index:13,09010
Boiling Pt:401 °C (1013 hPa)
Melting Pt:163…167 °C
Density:1.46 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:200-563-4
CAS Number:63-74-1
Formula:H₂NC₆H₄SO₂NH₂
MDL Number:MFCD00007939