
Sulphanilamide 98%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:63-74-1
:
Formula:H₂NC₆H₄SO₂NH₂
MDL Number:MFCD00007939
:172.21 g/mol
Merck Index:13,09010
Boiling Pt:401 °C (1013 hPa)
Melting Pt:163…167 °C
Density:1,46 g/cm³ (20 °C)
EINECS:200-563-4
:GHS07
Storage Temperature:Ambient
Specification Test Results
- Appearance:White to off-white Crystalline powder
- Infrared spectrum:Conforms
- Melting point:163 to 167 °C
- Loss on drying:≤0.5 % (1 g, 105 °C)
- HPLC:≥97.5 %