
3,4-Dimethoxyaniline, Sigma-Aldrich®
Catalog # SIALA83008-10G
Supplier: Merck
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:10 g
- Seq:3236
Specifications
About this item
:
Boiling Pt:296 °C (1013 hPa)
Melting Pt:85…88 °C
Storage Temperature:Ambient
EINECS:228-647-6
UN:2811
ADR:6.1,III
:M-Clarity™ quality level = MQ100
Flash Pt:>110 °C
:153.18 g/mol
:GHS07
CAS Number:6315-89-5
Formula:C₈H₁₁NO₂
MDL Number:MFCD00008394