1,4-Phenylene diisothiocyanate
Supplier: Thermo Scientific
Certificates
About this item
CAS Number:4044-65-9
:
Formula:C₈H₄N₂S₂
MDL Number:MFCD00004811
:192.27 g/mol
Merck Index:13,01304
Boiling Pt:319 °C (1013 hPa)
Melting Pt:131 °C
Storage Temperature:Refrigerator
EINECS:223-741-3
UN:2923
ADR:8,III