


2,4-Dimethoxyphenyl isocyanate ≥97%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:84370-87-6
:
Formula:C₉H₉NO₃
MDL Number:MFCD00002005
:179.18 g/mol
Boiling Pt:139…141 °C (11 mmHg)
Melting Pt:31…32 °C
Flash Pt:>110 °C (230 °F)
EINECS:282-747-4
UN:2811
ADR:6.1,III
:GHS06
:GHS08
:GHS07