4-Iodophenyl isocyanate, Sigma-Aldrich®
Glass bottle for solids, 5 g
Catalog # SIAL478687-5G
Supplier: Merck
4-Iodophenyl isocyanate, Sigma-Aldrich®
Catalog # SIAL478687-5G
Supplier: Merck
CAS Number: 15845-62-2
Jump to:
Product Details & Documents
Synonyms: 4-Iodophenylisocyanate
- CAS Number:15845-62-2
- Formula:C₇H₄INO
- MDL Number:MFCD00013875
- Molecular Weight:245.02 g/mol
- Boiling Pt:84 °C (5 mmHg)
- Melting Pt:45…46 °C
- Storage Temperature:Refrigerator
- UN:2811
- ADR:6.1,III
- :M-Clarity™ quality level = MQ200
Specifications
Specifications
- Pack type:Glass bottle for solids
- Pk:5 g



