2,4-Difluorothiophenol
Catalog # MAYBRF03663.1
Supplier: Thermo Scientific
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:146.16 g/mol
Boiling Pt:59 °C (1013 hPa)
Density:1.29 g/cm³ (20 °C)
UN:1993
ADR:3,III
CAS Number:1996-44-7
Formula:C₆H₄F₂S
MDL Number:MFCD00203477