2,4-Difluorophenol, Sigma-Aldrich®
Supplier: Merck
Certificates
About this item
CAS Number:367-27-1
:
Formula:C₆H₄F₂O
MDL Number:MFCD00009715
:130.09 g/mol
Boiling Pt:146 °C (1013 hPa)
Melting Pt:20 °C
Density:1.36 g/cm³ (20 °C)
EINECS:206-688-0
ADR:8,II
:M-Clarity™ quality level = MQ200
UN:2921