Order Entry
Export
Orders LinkContactUsLinkComponent
 

2,3-Difluorophenol, Sigma-Aldrich®

Glass bottle for solids, 5 g
Catalog # SIAL265322-5G
Supplier:   Merck
2,3-Difluorophenol, Sigma-Aldrich®
Catalog # SIAL265322-5G
Supplier:  Merck
CAS Number:  6418-38-8
2,3-Difluorophenol, Sigma-Aldrich®

Glass bottle for solids, 5 g

CAS Number:  6418-38-8
Price
47,90 €
Log in for availability
1 * 5 g
Product Details & Documents
  • CAS Number:6418-38-8
  • Formula:C₆H₄F₂O
  • MDL Number:MFCD00010262
  • Molecular Weight:130.09 g/mol
  • Boiling Pt:54 °C (25 mmHg)
  • Melting Pt:30…32 °C
  • UN:2921
  • ADR:8,II
  •  :M-Clarity™ quality level = MQ100
Specifications

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    5 g

Specifications