
1,3-Dimethoxy-5-fluorobenzene ≥98%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:52189-63-6
:
Formula:C₈H₉FO₂
MDL Number:MFCD00012445
:156.16 g/mol
Boiling Pt:86…88 °C (7 mmHg)
Density:1.170
Flash Pt:91 °C (196 °F)
:GHS07
Storage Temperature:Ambient