
1,4-Difluoro-2-nitrobenzene, Sigma-Aldrich®
Catalog # SIAL196622-25G
Supplier: Merck
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:25 g
- Seq:3308
Specifications
About this item
:
:159.09 g/mol
Boiling Pt:207 °C (1013 hPa)
Melting Pt:–11.7 °C
Density:1.4671 g/cm³ (17 °C)
EINECS:206-663-4
UN:2810
ADR:6.1,III
:M-Clarity™ quality level = MQ200
:GHS07
CAS Number:364-74-9
Formula:C₆H₃F₂NO₂
MDL Number:MFCD00007054