2-Fluoro-5-nitroaniline, Sigma-Aldrich®
Catalog # SIAL155853-25G
Supplier: Merck
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:25 g
- Seq:3308
Specifications
About this item
:
Melting Pt:99…101 °C
Storage Temperature:Ambient
EINECS:206-720-3
UN:1325
ADR:4.1,II
:M-Clarity™ quality level = MQ100
:156.12 g/mol
CAS Number:369-36-8
Formula:C₆H₅FN₂O₂
MDL Number:MFCD00007652