

2,6-Difluoroaniline, Sigma-Aldrich®
Supplier: Merck
Certificates
About this item
CAS Number:5509-65-9
:
Formula:C₆H₅F₂N
MDL Number:MFCD00007655
:129.11 g/mol
Boiling Pt:51 °C (1013 hPa)
Density:1.199 g/cm³ (20 °C)
EINECS:226-853-0
UN:1992
ADR:3,III
:M-Clarity™ quality level = MQ200
:GHS02
:GHS07
Storage Temperature:Ambient