2,3-Difluoroaniline, Sigma-Aldrich®
SIAL265306-5G
: Merck
:
- Pack type:Glass bottle for solids
- Pk:5 g
- Seq:3173
:
:129.11 g/mol
:69 °C (12 mmHg)
:1.274 g/cm³ (20 °C)
:6.1,III
:M-Clarity™ quality level = MQ100
:224-847-2
:2941
:4519-40-8
:C₆H₅F₂N
:MFCD00010298