
2,4-Difluorophenyl isothiocyanate
Supplier: Thermo Scientific
Certificates
About this item
CAS Number:141106-52-7
:
Formula:C₇H₃F₂NS
MDL Number:MFCD00041044
:171.17 g/mol
Boiling Pt:98…99 °C (30 mmHg)
Density:1.349 g/cm³ (20 °C)
UN:3334
ADR:9,
:GHS07