2,6-Difluorophenyl isothiocyanate
Catalog # MAYBAC30815.10
Supplier: Thermo Scientific
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:10 g
- Seq:3236
Specifications
Specification Test Results
- Appearance:White to light yellow Low melting solid or liquid
- Infrared spectrum:Conforms
- Refractive index:1.4945 to 1.4965 (20°C, 589 nm)
- GC:≥96.0 %
Specification Test Results
About this item
:
:171.17 g/mol
Boiling Pt:106…107 °C (35 mmHg)
Storage Temperature:Refrigerator
UN:2930
ADR:6.1,II
CAS Number:207974-17-2
Formula:C₇H₃F₂NS
MDL Number:MFCD00041047