2,5-Difluorophenyl isocyanate
Catalog # MAYBAC30914.10
Supplier: Thermo Scientific
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:10 g
- Seq:3236
Specifications
About this item
:
:155.1 g/mol
Boiling Pt:78 °C (33 mmHg)
Density:1.32 g/cm³ (20 °C)
Storage Temperature:Refrigerator
UN:3080
ADR:6.1,II
CAS Number:39718-32-6
Formula:C₇H₃F₂NO
MDL Number:MFCD00013843