Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
2,4-Difluorophenyl isocyanate
2,4-Difluorophenyl isocyanate, Sigma-Aldrich®
2,4-Difluorophenyl isocyanate, Sigma-Aldrich®
:
Merck
2,4-Difluorophenyl isocyanate, Sigma-Aldrich®
:
Merck
:
59025-55-7
:
:
C₇H₃F₂NO
:
MFCD00001997
:
42 °C (6 mmHg)
:
1.3 g/cm³ (20 °C)
:
6.1,II
:
M-Clarity™ quality level = MQ100
:
155.1 g/mol
:
Refrigerator
:
3080