2,4-Difluorophenyl isocyanate
Supplier: Thermo Scientific
Certificates
About this item
CAS Number:59025-55-7
:
Formula:C₇H₃F₂NO
MDL Number:MFCD00001997
Boiling Pt:42 °C (6 mmHg)
Density:1.3 g/cm³ (20 °C)
Storage Temperature:Refrigerator
UN:3080
ADR:6.1,II
:155.1 g/mol