Order Entry
Export
ContactUsLinkComponent
2,4,5-Trifluoro-3-methoxybenzoic acid
2,4,5-Trifluoro-3-methoxybenzoic acid
Catalog # MOLE55476475-1G
Supplier:  Molekula
CAS Number:  
2,4,5-Trifluoro-3-methoxybenzoic acid
Catalog # MOLE55476475-1G
Supplier:  Molekula
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :206.12 g/mol
CAS Number:112811-65-1
Formula:C₈H₅F₃O₃
MDL Number:MFCD00153201