
3,4-Difluorobenzoic acid, Sigma-Aldrich®
Catalog # SIAL264318-5G
Supplier: Merck
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:5 g
- Seq:3173
Specifications
About this item
:
:158.1 g/mol
Boiling Pt:242 °C (1013 hPa)
Melting Pt:119…122 °C
Density:1 g/cm³ (20 °C)
:M-Clarity™ quality level = MQ100
Storage Temperature:Ambient
EINECS:207-249-6
:GHS07
CAS Number:455-86-7
Formula:C₇H₄F₂O₂
MDL Number:MFCD00011672